Assay Details
| Gene Symbol: IKBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244129.1 | NP_001231058.1 | 2 - 3 | 663 | 98 | |
| XM_005664219.3 | XP_005664276.1 | 438 | 98 | |||
| XM_013998047.2 | XP_013853501.1 | 530 | 98 | |||
| GenBank mRNA | AK348922.1 | - | 2 - 3 | 663 | 98 | |
More Information
Set Membership: |
|

IKBIP
Genomic Map