Assay Details
| Gene Symbol: SSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244147.1 | NP_001231076.1 | 4 - 5 | 588 | 80 | |
| GenBank mRNA | AK231884.1 | - | 4 - 5 | 590 | 80 | |
| AK232862.1 | - | 4 - 5 | 588 | 80 | ||
| AK345822.1 | - | 4 - 5 | 554 | 80 | ||
| AK350259.1 | - | 4 - 5 | 553 | 80 | ||
| AY610158.1 | - | 4 - 5 | 562 | 80 | ||
More Information
Set Membership: |
|

SSR3
Genomic Map