Assay Details
| Gene Symbol: COPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244149.1 | NP_001231078.1 | 10 - 11 | 1225 | 56 | |
| XM_021066089.1 | XP_020921748.1 | 1099 | 56 | |||
| GenBank mRNA | AK234845.1 | - | 9 - 10 | 996 | 56 | |
| AK239091.1 | - | 10 - 11 | 1221 | 56 | ||
| AK395140.1 | - | 9 - 10 | 1082 | 56 | ||
| AK396275.1 | - | 10 - 11 | 1497 | 56 | ||
| CK459694.1 | - | 7 - 8 | 593 | 56 | ||
More Information
Set Membership: |
|

COPS3
Genomic Map