Assay Details
| Gene Symbol: FKBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244157.2 | NP_001231086.1 | 6 - 7 | 686 | 96 | |
| GenBank mRNA | AK239153.1 | - | 6 - 7 | 646 | 96 | |
| AK240163.1 | - | 6 - 7 | 641 | 96 | ||
| AK352247.1 | - | 6 - 7 | 641 | 96 | ||
More Information
Set Membership: |
|

FKBP3
Genomic Map