Assay Details
| Gene Symbol: MTCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244166.1 | NP_001231095.1 | 2 - 3 | 444 | 62 | |
| NM_001244167.1 | NP_001231096.1 | 2 - 3 | 444 | 62 | ||
| XM_021097989.1 | XP_020953648.1 | 438 | 62 | |||
| XM_021097990.1 | XP_020953649.1 | 419 | 62 | |||
| GenBank mRNA | AK231630.1 | - | 2 - 3 | 423 | 62 | |
| AK231935.1 | - | 2 - 3 | 444 | 62 | ||
| AK345684.1 | - | 2 - 3 | 407 | 62 | ||
| AK351214.1 | - | 2 - 3 | 391 | 62 | ||
| AK392530.1 | - | 2 - 3 | 439 | 62 | ||
| AK392692.1 | - | 2 - 3 | 450 | 62 | ||
| AK396897.1 | - | 2 - 3 | 438 | 62 | ||
| AK396911.1 | - | 2 - 3 | 387 | 62 | ||
| AK397268.1 | - | 2 - 3 | 441 | 62 | ||
| BP439986.1 | - | 2 - 3 | 444 | 62 | ||
More Information
Set Membership: |
|

MTCH1
Genomic Map