Assay Details
| Gene Symbol: PDE6H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244244.1 | NP_001231173.1 | 2 - 3 | 172 | 98 | |
| XM_021090955.1 | XP_020946614.1 | 391 | 98 | |||
| XM_021090956.1 | XP_020946615.1 | 544 | 98 | |||
| GenBank mRNA | EW533113.2 | - | 2 - 3 | 67 | 98 | |
More Information
Set Membership: |
|

PDE6H
Genomic Map