Assay Details
| Gene Symbol: SLC33A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244245.1 | NP_001231174.1 | 5 - 6 | 1907 | 69 | |
| XM_005669929.3 | XP_005669986.1 | 1939 | 69 | |||
| XM_021068489.1 | XP_020924148.1 | 1989 | 69 | |||
| GenBank mRNA | AK349530.1 | - | 5 - 6 | 1906 | 69 | |
| AK400803.1 | - | 5 - 6 | 1907 | 69 | ||
More Information
Set Membership: |
|

SLC33A1
Genomic Map