Assay Details
| Gene Symbol: NOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244246.1 | NP_001231175.1 | 5 - 6 | 583 | 60 | |
| XM_013987297.2 | XP_013842751.1 | 674 | 60 | |||
| GenBank mRNA | AK343646.1 | - | 5 - 6 | 562 | 60 | |
| AK343669.1 | - | 5 - 6 | 562 | 60 | ||
| AK343749.1 | - | 5 - 6 | 569 | 60 | ||
| AK345763.1 | - | 5 - 6 | 584 | 60 | ||
| AK391033.1 | - | 5 - 6 | 568 | 60 | ||
| AK393604.1 | - | 5 - 6 | 582 | 60 | ||
More Information
Set Membership: |
|

NOP2
Genomic Map