Assay Details
| Gene Symbol: POGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244259.1 | NP_001231188.1 | 4 - 5 | 1840 | 68 | |
| XM_021088566.1 | XP_020944225.1 | 2079 | 68 | |||
| XM_021088567.1 | XP_020944226.1 | 1957 | 68 | |||
| XM_021088568.1 | XP_020944227.1 | 1826 | 68 | |||
| XM_021088569.1 | XP_020944228.1 | 1607 | 68 | |||
| GenBank mRNA | AK395673.1 | - | 5 - 6 | 1894 | 68 | |
| EU814891.1 | - | 4 - 5 | 1840 | 68 | ||
More Information
Set Membership: |
|

POGK
Genomic Map