Assay Details
| Gene Symbol: TXNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244276.1 | NP_001231205.1 | 3 - 4 | 569 | 81 | |
| XM_013985373.2 | XP_013840827.1 | 630 | 81 | |||
| GenBank mRNA | AK232713.1 | - | 3 - 4 | 569 | 81 | |
| AK234889.1 | - | 3 - 4 | 599 | 81 | ||
| AK393071.1 | - | 3 - 4 | 659 | 81 | ||
| AY609590.1 | - | 3 - 4 | 626 | 81 | ||
More Information
Set Membership: |
|

TXNL1
Genomic Map