Assay Details
Gene Symbol: CD300C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244303.1 | NP_001231232.1 | 2 - 3 | 778 | 63 | |
GenBank mRNA | AJ949638.1 | - | 2 - 3 | 483 | 63 | |
AJ951325.1 | - | 1 - 2 | 234 | 63 | ||
AK395956.1 | - | 2 - 3 | 757 | 63 | ||
CJ020062.1 | - | 2 - 3 | 750 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
