Assay Details
| Gene Symbol: NAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244316.1 | NP_001231245.1 | 11 - 12 | 1054 | 75 | |
| NM_001244317.1 | NP_001231246.1 | 11 - 12 | 1054 | 75 | ||
| XM_005663991.3 | XP_005664048.1 | 1102 | 75 | |||
| XM_005663993.3 | XP_005664050.1 | 904 | 75 | |||
| XM_021091082.1 | XP_020946741.1 | 1017 | 75 | |||
| XM_021091083.1 | XP_020946742.1 | 904 | 75 | |||
| GenBank mRNA | AK233368.1 | - | 11 - 12 | 1054 | 75 | |
| AK236421.1 | - | 10 - 11 | 1020 | 75 | ||
| AK238821.1 | - | 9 - 10 | 866 | 75 | ||
| AK389439.1 | - | 11 - 12 | 1083 | 75 | ||
| AK393220.1 | - | 11 - 12 | 1055 | 75 | ||
| AK395665.1 | - | 11 - 12 | 1057 | 75 | ||
| AK398816.1 | - | 11 - 12 | 1057 | 75 | ||
| AY610037.2 | - | 11 - 12 | 1038 | 75 | ||
More Information
Set Membership: |
|

NAP1L1
Genomic Map