Assay Details
| Gene Symbol: PHF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244327.1 | NP_001231256.1 | 8 - 9 | 756 | 88 | |
| XM_005668398.3 | XP_005668455.1 | 780 | 88 | |||
| GenBank mRNA | AK235737.1 | - | 8 - 9 | 686 | 88 | |
| AK239065.1 | - | 8 - 9 | 749 | 88 | ||
| AK395116.1 | - | 8 - 9 | 754 | 88 | ||
| AK399378.1 | - | 5 - 6 | 393 | 88 | ||
| BW982187.1 | - | 8 - 9 | 749 | 88 | ||
More Information
Set Membership: |
|

PHF11
Genomic Map