Assay Details
| Gene Symbol: PHF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244327.1 | NP_001231256.1 | 9 - 10 | 819 | 78 | |
| XM_005668398.3 | XP_005668455.1 | 843 | 78 | |||
| XM_013980495.2 | XP_013835949.1 | 775 | 78 | |||
| GenBank mRNA | AK235737.1 | - | 9 - 10 | 749 | 78 | |
| AK239065.1 | - | 9 - 10 | 812 | 78 | ||
| AK395116.1 | - | 9 - 10 | 78 | |||
| AK399378.1 | - | 6 - 7 | 456 | 78 | ||
More Information
Set Membership: |
|

PHF11
Genomic Map