Assay Details
| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244334.1 | NP_001231263.1 | 2 - 3 | 352 | 70 | |
| XM_021093319.1 | XP_020948978.1 | 1136 | 70 | |||
| XM_021093320.1 | XP_020948979.1 | 1260 | 70 | |||
| GenBank mRNA | AJ684804.1 | - | 2 - 3 | 319 | 70 | |
| AK349407.1 | - | 2 - 3 | 352 | 70 | ||
| EW468677.2 | - | 1 - 2 | 169 | 70 | ||
More Information
Set Membership: |
|

CIBAR2
Genomic Map