Assay Details
| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244334.1 | NP_001231263.1 | 3 - 4 | 426 | 79 | |
| XM_021093319.1 | XP_020948978.1 | 1210 | 79 | |||
| XM_021093320.1 | XP_020948979.1 | 1334 | 79 | |||
| GenBank mRNA | AK349407.1 | - | 3 - 4 | 426 | 79 | |
| EW468677.2 | - | 2 - 3 | 243 | 79 | ||
More Information
Set Membership: |
|

CIBAR2
Genomic Map