Assay Details
| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244334.1 | NP_001231263.1 | 8 - 9 | 847 | 92 | |
| GenBank mRNA | AK349407.1 | - | 8 - 9 | 847 | 92 | |
| EW061678.2 | - | 1 - 2 | 142 | 92 | ||
More Information
Set Membership: |
|

CIBAR2
Genomic Map