Assay Details
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244335.1 | NP_001231264.1 | 2 - 3 | 839 | 92 | |
GenBank mRNA | AK240506.1 | - | 6 - 7 | 1292 | 92 | |
AK352171.1 | - | 7 - 8 | 1700 | 92 | ||
AK391698.1 | - | 2 - 3 | 700 | 92 | ||
AK392453.1 | - | 2 - 3 | 788 | 92 | ||
AK399915.1 | - | 2 - 3 | 747 | 92 | ||
AK400150.1 | - | 2 - 3 | 801 | 92 | ||
FS666793.1 | - | 2 - 3 | 715 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
