Assay Details
| Gene Symbol: ADPRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244347.1 | NP_001231276.1 | 3 - 4 | 721 | 82 | |
| XM_021068214.1 | XP_020923873.1 | 1088 | 82 | |||
| XM_021068215.1 | XP_020923874.1 | 598 | 82 | |||
| XM_021068216.1 | XP_020923875.1 | 849 | 82 | |||
| XM_021068217.1 | XP_020923876.1 | 1088 | 82 | |||
| GenBank mRNA | AK236200.1 | - | 3 - 4 | 723 | 82 | |
| AK236359.1 | - | 3 - 4 | 721 | 82 | ||
| AK240388.1 | - | 3 - 4 | 559 | 82 | ||
| AK391059.1 | - | 3 - 4 | 796 | 82 | ||
| AK398702.1 | - | 3 - 4 | 560 | 82 | ||
More Information
Set Membership: |
|

ADPRH
Genomic Map