Assay Details
| Gene Symbol: EIF1AY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244349.1 | NP_001231278.1 | 6 - 7 | 547 | 139 | |
| XM_013991067.2 | XP_013846521.2 | 543 | 139 | |||
| GenBank mRNA | AY609402.1 | - | 6 - 7 | 547 | 139 | |
More Information
Set Membership: |
|

EIF1AY
Genomic Map