Assay Details
| Gene Symbol: IFIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244363.1 | NP_001231292.1 | 1 - 2 | 214 | 106 | |
| GenBank mRNA | AK230745.1 | - | 1 - 2 | 71 | 106 | |
| AK230927.1 | - | 1 - 2 | 67 | 106 | ||
| AK236882.1 | - | 1 - 2 | 98 | 106 | ||
| AK343760.1 | - | 1 - 2 | 72 | 106 | ||
| AK343860.1 | - | 1 - 2 | 106 | |||
| AK399100.1 | - | 1 - 2 | 94 | 106 | ||
| HQ679904.1 | - | 1 - 2 | 48 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244363.1 transcripts.
Set Membership: |
|

IFIT1
Genomic Map