Assay Details
Gene Symbol: PRXL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244373.2 | NP_001231302.1 | 5 - 6 | 729 | 109 | |
NM_001244374.2 | NP_001231303.1 | 4 - 5 | 615 | 109 | ||
NM_001244375.2 | NP_001231304.1 | 4 - 5 | 598 | 109 | ||
NM_001359936.1 | NP_001346865.1 | 5 - 6 | 712 | 109 | ||
NM_001359937.1 | NP_001346866.1 | 4 - 5 | 448 | 109 | ||
NM_001359938.1 | NP_001346867.1 | 4 - 5 | 465 | 109 | ||
NM_001359939.1 | NP_001346868.1 | 5 - 6 | 595 | 109 | ||
XM_005671141.3 | XP_005671198.1 | 526 | 109 | |||
GenBank mRNA | AK232037.1 | - | 5 - 6 | 646 | 109 | |
AK234200.1 | - | 4 - 5 | 533 | 109 | ||
AK389174.1 | - | 4 - 5 | 558 | 109 | ||
AK400395.1 | - | 5 - 6 | 712 | 109 | ||
AY610040.1 | - | 3 - 4 | 666 | 109 | ||
DB781655.1 | - | 4 - 5 | 558 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
