Assay Details
| Gene Symbol: PTPMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244377.1 | NP_001231306.1 | 3 - 4 | 645 | 91 | |
| GenBank mRNA | AK344846.1 | - | 2 - 3 | 360 | 91 | |
| EW202464.2 | - | 1 - 2 | 45 | 91 | ||
| EW394041.2 | - | 3 - 4 | 447 | 91 | ||
| EW493122.2 | - | 3 - 4 | 550 | 91 | ||
More Information
Set Membership: |
|

PTPMT1
Genomic Map