Assay Details
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244382.2 | NP_001231311.1 | 5 - 6 | 736 | 76 | |
NM_001244383.2 | NP_001231312.2 | 5 - 6 | 665 | 76 | ||
XM_005671150.3 | XP_005671207.1 | 611 | 76 | |||
XM_005671151.3 | XP_005671208.1 | 686 | 76 | |||
XM_005671152.3 | XP_005671209.1 | 788 | 76 | |||
GenBank mRNA | AK239027.1 | - | 5 - 6 | 551 | 76 | |
AK239590.1 | - | 5 - 6 | 638 | 76 | ||
AK239697.1 | - | 5 - 6 | 597 | 76 | ||
AK389707.1 | - | 5 - 6 | 639 | 76 | ||
AK392804.1 | - | 5 - 6 | 687 | 76 | ||
AK395192.1 | - | 5 - 6 | 682 | 76 | ||
AK398770.1 | - | 5 - 6 | 718 | 76 | ||
AY610005.1 | - | 5 - 6 | 615 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
