Assay Details
| Gene Symbol: CDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244385.1 | NP_001231314.1 | 2 - 3 | 323 | 103 | |
| GenBank mRNA | AK346675.1 | - | 2 - 3 | 301 | 103 | |
| AY610472.1 | - | 2 - 3 | 323 | 103 | ||
More Information
Set Membership: |
|

CDA
Genomic Map