Assay Details
| Gene Symbol: SBDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244393.1 | NP_001231322.1 | 4 - 5 | 856 | 78 | |
| GenBank mRNA | AK396822.1 | - | 4 - 5 | 886 | 78 | |
| AY609615.1 | - | 4 - 5 | 856 | 78 | ||
More Information
Set Membership: |
|

SBDS
Genomic Map