Assay Details
| Gene Symbol: RABEPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244400.1 | NP_001231329.1 | 4 - 5 | 437 | 70 | |
| XM_005654539.3 | XP_005654596.1 | 784 | 70 | |||
| XM_005654541.3 | XP_005654598.1 | 500 | 70 | |||
| XM_013986587.2 | XP_013842041.1 | 678 | 70 | |||
| XM_013986596.2 | XP_013842050.1 | 419 | 70 | |||
| GenBank mRNA | AK240368.1 | - | 4 - 5 | 437 | 70 | |
| AK389388.1 | - | 4 - 5 | 437 | 70 | ||
More Information
Set Membership: |
|

RABEPK
Genomic Map