Assay Details
| Gene Symbol: LCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244410.1 | NP_001231339.1 | 2 - 3 | 473 | 84 | |
| GenBank mRNA | AK240091.1 | - | 2 - 3 | 473 | 84 | |
| AK352158.1 | - | 2 - 3 | 333 | 84 | ||
| FM995245.1 | - | 1 - 2 | 50 | 84 | ||
More Information
Set Membership: |
|

LCN2
Genomic Map