Assay Details
| Gene Symbol: LCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244410.1 | NP_001231339.1 | 3 - 4 | 548 | 62 | |
| XM_021088538.1 | XP_020944197.1 | 533 | 62 | |||
| GenBank mRNA | AK240091.1 | - | 3 - 4 | 548 | 62 | |
| AK352158.1 | - | 3 - 4 | 408 | 62 | ||
More Information
Set Membership: |
|

LCN2
Genomic Map