Assay Details
| Gene Symbol: SLC1A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244425.2 | NP_001231354.1 | 7 - 8 | 1312 | 70 | |
| NM_001289944.1 | NP_001276873.1 | 6 - 7 | 1174 | 70 | ||
| NM_001289945.1 | NP_001276874.1 | 7 - 8 | 1312 | 70 | ||
| NM_001289946.1 | NP_001276875.1 | 5 - 6 | 976 | 70 | ||
| XM_021076550.1 | XP_020932209.1 | 2954 | 70 | |||
| GenBank mRNA | AK231079.1 | - | 7 - 8 | 1301 | 70 | |
| AK389716.1 | - | 7 - 8 | 1312 | 70 | ||
| HQ699552.1 | - | 4 - 5 | 758 | 70 | ||
| HQ699553.1 | - | 6 - 7 | 1094 | 70 | ||
| HQ699554.1 | - | 6 - 7 | 1094 | 70 | ||
More Information
Set Membership: |
|

SLC1A3
Genomic Map