Assay Details
| Gene Symbol: SLC31A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244427.1 | NP_001231356.1 | 3 - 4 | 371 | 72 | |
| XM_013993750.2 | XP_013849204.1 | 1228 | 72 | |||
| GenBank mRNA | AK232136.1 | - | 3 - 4 | 364 | 72 | |
| AK350457.1 | - | 3 - 4 | 371 | 72 | ||
More Information
Set Membership: |
|

SLC31A2
Genomic Map