Assay Details
| Gene Symbol: CLIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244428.1 | NP_001231357.1 | 2 - 3 | 229 | 100 | |
| XM_021079470.1 | XP_020935129.1 | 245 | 100 | |||
| GenBank mRNA | AK230814.1 | - | 2 - 3 | 229 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021079470.1 transcripts.
Set Membership: |
|

CLIC2
Genomic Map