Assay Details
| Gene Symbol: CLIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244428.1 | NP_001231357.1 | 3 - 4 | 351 | 84 | |
| XM_021079470.1 | XP_020935129.1 | 367 | 84 | |||
| GenBank mRNA | AK230814.1 | - | 3 - 4 | 351 | 84 | |
More Information
Set Membership: |
|

CLIC2
Genomic Map