Assay Details
| Gene Symbol: GAL3ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244429.1 | NP_001231358.1 | 2 - 3 | 433 | 107 | |
| XM_005670827.2 | XP_005670884.1 | 259 | 107 | |||
| GenBank mRNA | AK231249.1 | - | 2 - 3 | 433 | 107 | |
More Information
Set Membership: |
|

GAL3ST1
Genomic Map