Assay Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244441.1 | NP_001231370.1 | 2 - 3 | 337 | 79 | |
| GenBank mRNA | AW358764.1 | - | 2 - 3 | 95 | 79 | |
More Information
Set Membership: |
|

FAM216A
Genomic Map