Assay Details
| Gene Symbol: RNASEH2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244444.1 | NP_001231373.1 | 4 - 5 | 513 | 72 | |
| XM_005652860.3 | XP_005652917.1 | 1228 | 72 | |||
| XM_013990663.2 | XP_013846117.1 | 1527 | 72 | |||
| XM_013990670.2 | XP_013846124.1 | 1434 | 72 | |||
| XM_013990688.2 | XP_013846142.1 | 2040 | 72 | |||
| XM_021079423.1 | XP_020935082.1 | 1563 | 72 | |||
| XM_021079430.1 | XP_020935089.1 | 1220 | 72 | |||
| XM_021079433.1 | XP_020935092.1 | 1048 | 72 | |||
| XM_021079434.1 | XP_020935093.1 | 1064 | 72 | |||
| XM_021079437.1 | XP_020935096.1 | 1498 | 72 | |||
| XM_021079439.1 | XP_020935098.1 | 1253 | 72 | |||
| XM_021079456.1 | XP_020935115.1 | 1064 | 72 | |||
| GenBank mRNA | AK343438.1 | - | 4 - 5 | 513 | 72 | |
More Information
Set Membership: |
|

RNASEH2A
Genomic Map