Assay Details
| Gene Symbol: NME3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244449.1 | NP_001231378.1 | 3 - 4 | 305 | 75 | |
| NM_001244450.1 | NP_001231379.1 | 3 - 4 | 302 | 75 | ||
| NR_045137.1 | 2 - 3 | 357 | 75 | |||
| XM_013987752.2 | XP_013843206.1 | 489 | 75 | |||
| XM_013987753.2 | XP_013843207.1 | 486 | 75 | |||
| GenBank mRNA | AK240056.1 | - | 2 - 3 | 353 | 75 | |
| AK394553.1 | - | 2 - 3 | 356 | 75 | ||
| BF080492.1 | - | 3 - 4 | 305 | 75 | ||
| BW980049.1 | - | 3 - 4 | 274 | 75 | ||
| CJ014819.1 | - | 2 - 3 | 357 | 75 | ||
More Information
Set Membership: |
|

NME3
Genomic Map