Assay Details
| Gene Symbol: PSMB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244455.1 | NP_001231384.1 | 3 - 4 | 537 | 59 | |
| GenBank mRNA | AK234102.1 | - | 3 - 4 | 524 | 59 | |
| AK235425.1 | - | 3 - 4 | 515 | 59 | ||
| AK239491.1 | - | 3 - 4 | 518 | 59 | ||
| AK240608.1 | - | 3 - 4 | 515 | 59 | ||
| AK348625.1 | - | 3 - 4 | 520 | 59 | ||
| AK393768.1 | - | 3 - 4 | 531 | 59 | ||
| AY609648.1 | - | 3 - 4 | 532 | 59 | ||
| FS667250.1 | - | 3 - 4 | 538 | 59 | ||
More Information
Set Membership: |
|

PSMB4
Genomic Map