Assay Details
| Gene Symbol: ERG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244456.1 | NP_001231385.1 | 8 - 9 | 988 | 72 | |
| XM_021068468.1 | XP_020924127.1 | 1156 | 72 | |||
| XM_021068469.1 | XP_020924128.1 | 1084 | 72 | |||
| XM_021068470.1 | XP_020924129.1 | 935 | 72 | |||
| XM_021068471.1 | XP_020924130.1 | 1003 | 72 | |||
| XM_021068472.1 | XP_020924131.1 | 854 | 72 | |||
| GenBank mRNA | AK234482.1 | - | 8 - 9 | 988 | 72 | |
| AK236690.1 | - | 2 - 3 | 122 | 72 | ||
| AK395618.1 | - | 6 - 7 | 799 | 72 | ||
More Information
Set Membership: |
|

ERG
Genomic Map