Assay Details
| Gene Symbol: PRDX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244474.1 | NP_001231403.1 | 1 - 2 | 110 | 77 | |
| GenBank mRNA | AK349436.1 | - | 1 - 2 | 110 | 77 | |
| AK390553.1 | - | 1 - 2 | 59 | 77 | ||
| AK392270.1 | - | 1 - 2 | 78 | 77 | ||
| AK400020.1 | - | 1 - 2 | 110 | 77 | ||
| GQ913659.1 | - | 1 - 2 | 64 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244474.1 transcripts.
Set Membership: |
|

PRDX2
Genomic Map