Assay Details
| Gene Symbol: NFATC2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244522.1 | NP_001231451.1 | 5 - 6 | 858 | 59 | |
| XM_005662047.3 | XP_005662104.2 | 943 | 59 | |||
| GenBank mRNA | AK235025.1 | - | 5 - 6 | 858 | 59 | |
| AK395742.1 | - | 5 - 6 | 862 | 59 | ||
More Information
Set Membership: |
|

NFATC2IP
Genomic Map