Assay Details
| Gene Symbol: FKBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244527.1 | NP_001231456.1 | 1 - 2 | 371 | 96 | |
| GenBank mRNA | EW372471.2 | - | 1 - 2 | 365 | 96 | |
| EW454624.2 | - | 1 - 2 | 212 | 96 | ||
| FD642407.1 | - | 1 - 2 | 371 | 96 | ||
More Information
Set Membership: |
|

FKBP14
Genomic Map