Assay Details
| Gene Symbol: DDX56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244543.1 | NP_001231472.1 | 12 - 13 | 1526 | 57 | |
| GenBank mRNA | AK234104.1 | - | 12 - 13 | 1526 | 57 | |
| AK234966.1 | - | 10 - 11 | 1146 | 57 | ||
| AK351264.1 | - | 10 - 11 | 1106 | 57 | ||
| AK392383.1 | - | 12 - 13 | 2109 | 57 | ||
| AK396423.1 | - | 12 - 13 | 1504 | 57 | ||
More Information
Set Membership: |
|

DDX56
Genomic Map