Assay Details
| Gene Symbol: ARPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244544.1 | NP_001231473.1 | 2 - 3 | 264 | 63 | |
| XM_021098270.1 | XP_020953929.1 | 394 | 63 | |||
| GenBank mRNA | AK348002.1 | - | 2 - 3 | 264 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021098270.1 transcripts.
Set Membership: |
|

ARPIN
Genomic Map