Assay Details
| Gene Symbol: MRPS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244553.1 | NP_001231482.1 | 10 - 11 | 1127 | 73 | |
| XM_021085713.1 | XP_020941372.1 | 1313 | 73 | |||
| GenBank mRNA | AK239394.1 | - | 10 - 11 | 1127 | 73 | |
| AK390436.1 | - | 10 - 11 | 1132 | 73 | ||
More Information
Set Membership: |
|

MRPS9
Genomic Map