Assay Details
| Gene Symbol: TNFSF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244555.1 | NP_001231484.1 | 3 - 4 | 363 | 65 | |
| GenBank mRNA | AK345356.1 | - | 3 - 4 | 363 | 65 | |
More Information
Set Membership: |
|

TNFSF15
Genomic Map