Assay Details
| Gene Symbol: NSF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244560.1 | NP_001231489.1 | 12 - 13 | 1398 | 67 | |
| XM_005668696.3 | XP_005668753.1 | 1515 | 67 | |||
| XM_013980864.2 | XP_013836318.1 | 1466 | 67 | |||
| GenBank mRNA | AK230846.1 | - | 12 - 13 | 1398 | 67 | |
| AK391859.1 | - | 12 - 13 | 1504 | 67 | ||
| AK399551.1 | - | 12 - 13 | 1415 | 67 | ||
More Information
Set Membership: |
|

NSF
Genomic Map