Assay Details
| Gene Symbol: SLC24A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244595.1 | NP_001231524.1 | 5 - 6 | 1287 | 72 | |
| XM_021078223.1 | XP_020933882.1 | 1571 | 72 | |||
| XM_021078277.1 | XP_020933936.1 | 1491 | 72 | |||
| XM_021078320.1 | XP_020933979.1 | 1512 | 72 | |||
| XM_021078387.1 | XP_020934046.1 | 1451 | 72 | |||
More Information
Set Membership: |
|

SLC24A2
Genomic Map