Assay Details
| Gene Symbol: C7H6orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244686.1 | NP_001231615.1 | 1 | 814 | 81 | |
| GenBank mRNA | AK237470.1 | - | 1 | 815 | 81 | |
| AK350319.1 | - | 1 | 581 | 81 | ||
More Information
Set Membership: |
|

C7H6orf47
Genomic Map