Assay Details
| Gene Symbol: RPP21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244694.1 | NP_001231623.1 | 2 - 3 | 237 | 97 | |
| XM_005653456.2 | XP_005653513.1 | 213 | 97 | |||
| GenBank mRNA | AK399671.1 | - | 2 - 3 | 232 | 97 | |
| EW455534.2 | - | 2 - 3 | 186 | 97 | ||
More Information
Set Membership: |
|

RPP21
Genomic Map